HUASMC, Cryopreserved
HUASMC, Cryopreserved
HUASMC, cryopreserved, 0,5M cells (5×10⁵ cells/vial), Cryopreserved, Single donor, 1 Vial
Catalog # PRTV1210611
Supplier:   PROVITRO
HUASMC, Cryopreserved
Catalog # PRTV1210611
Supplier:  PROVITRO
CAS Number:  
HUASMC, Cryopreserved
HUASMC, Cryopreserved
HUASMC, cryopreserved, 0,5M cells (5×10⁵ cells/vial), Cryopreserved, Single donor, 1 Vial
CAS Number:  
Price
802,00 €
Log in for availability
1 * 1 VIA
Product Details & Documents
HUASMC - Human umbilical artery smooth muscle cells - are obtained from human tissue, in particularly from the umbilical artery, initially isolated in their in vivo state. The cells are then transitioned into a primary cell culture and subsequently aliquoted and frozen for preservation. It's crucial to note that HUASMC cultures undergo careful methods during isolation, ensuring that they remain untransformed and unmutated, maintaining characteristics reflective of their in vivo state. These primary cultures have a limited in-vitro lifespan.
  • It's crucial to note that HUASMC cultures undergo careful methods during isolation, ensuring that they remain untransformed and unmutated, maintaining characteristics reflective of their in vivo state. These primary cultures have a limited in-vitro lifespan.
  • Each batch of HUASMC undergoes rigorous testing, specifically assessing their proliferative capacity. These assessments are conducted using provitro's culture reagents, as detailed in the accompanying certificate of analysis. This stringent testing process ensures the consistency and reliability of HUASMC cultures for research and experimental applications
Each batch of HUASMC undergoes rigorous testing, specifically assessing their proliferative capacity. These assessments are conducted using provitro's culture reagents, as detailed in the accompanying certificate of analysis. This stringent testing process ensures the consistency and reliability of HUASMC cultures for research and experimental applications. This product contain 500,000 cells/cryovial.
Storge: Check the cryovial for signs of damage upon receipt. Since the cryopreserved HUASMC samples are shipped on dry ice, they must be immediately transferred to a freezer at −80 °C for storage, either for a period of up to one week or after the thawing process described below. In cases where longer storage is required, the vial should be transferred directly to liquid nitrogen at a temperature of −196 °C. This ensures optimal preservation of the vial. This ensures optimal preservation of the cryopreserved HUASMC samples for longer-term storage requirements.
Storge: Check the cryovial for signs of damage upon receipt. Since the cryopreserved HUASMC samples are shipped on dry ice, they must be immediately transferred to a freezer at −80 °C for storage, either for a period of up to one week or after the thawing process described below. In cases where longer storage is required, the vial should be transferred directly to liquid nitrogen at a temperature of −196 °C. This ensures optimal preservation of the vial. This ensures optimal preservation of the cryopreserved HUASMC samples for longer-term storage requirements.
Specifications
Specifications
  • Format de cellule:
    Cryopreserved
  • Cdt:
    1 Vial
  • Donneur unique/donneurs regroupés:
    Single donor
  • Désignation:
    HUASMC, cryopreserved, 0,5M cells (5×10⁵ cells/vial)