Water Estimator, Dean-Stark
Dean-Stark water estimator, gauge tube, xylene method, sleeve and core, 10 ml, 1,0 ml, NS 29/32 NS
N° de catalogue NEUB0381-04-101292
Fournisseur: NEUBERT VOLUME GLASSWAERE
Water Estimator, Dean-Stark
N° de catalogue NEUB0381-04-101292
Fournisseur: NEUBERT VOLUME GLASSWAERE
Aller à :
Product Details & Documents
Water estimator tubes for circling water with specific lighter solvents like toluene, benzene, cyclohexane.
Gauge tube - without tap, single part, xylene method, 0-point at the bottom; according to DIN 12420, with short-conical base.
Spécifications
- Rodage femelle:NS 29/32 NS
- Capacité:10 ml
- Désignation:Dean-Stark water estimator, gauge tube, xylene method, sleeve and core
- Division échelle:1,0 ml