Specifications
- Désignation:EasySep™ release mouse biotin positive selection kit, 1×10⁹ cells
- Cdt:1 KIT
Specifications
About this item
Immunomagnetic positive selection of particle-free mouse cells labelled with biotinylated antibodies.
- Highly purified cells labelled with biotinylated antibodies isolated from mouse tissue in less than 40 minutes
- No-wash removal of EasySep™ Releasable RapidSpheres™
The EasySep™ Release Mouse Biotin Positive Selection Kit is designed to isolate cells that are labelled with biotin-conjugated antibodies from mouse tissue by immunomagnetic positive selection.
Desired cells are labelled with antibodies and magnetic particles, and separated without columns using an EasySep™ magnet. Unwanted cells are simply poured off, while desired cells remain in the tube. Then, bound magnetic particles are removed from the EasySep™-isolated, biotin-antibody labelled cells, which are immediately available for downstream applications. Following cell isolation with this EasySep™ Release kit, antibody complexes remain bound to the cell surface and may interact with Brilliant Violet™ antibody conjugates, polyethylene glycol-modified proteins, or other chemically related ligands. This product replaces the EasySep™ Mouse Biotin Positive Selection Kit, providing highly purified particle-free cells.