EasySep™ Mouse Pan-ILC Enrichment Kit
Catalog # STMC19875
Supplier: STEMCELL Technologies
Specifications
- Désignation:EasySep™ mouse pan-ILC enrichment kit, 1×10⁹ cells
- Cdt:1 KIT
Specifications
About this item
Immunomagnetic enrichment of untouched mouse ILC1, ILC2, and ILC3s
- Fast, easy-to-use and column-free
- Isolated cells are untouched
- Facilitates rapid flow sorting of ILCs
This product is designed to enrich group 1, 2 and 3 innate lymphoid cells (ILC1, ILC2 and ILC3s) from single-cell suspensions of mouse tissues by negative selection. Unwanted cells are targeted for removal with biotinylated antibodies directed against non-ILCs and streptavidin-coated magnetic particles (RapidSpheres™). labelled cells are separated using an EasySep™ magnet without the use of columns. Unwanted cells remain in the tube as desired cells are simply poured off into a new tube. Isolated cells are immediately available for downstream applications such as flow cytometry and cell sorting.
Caution: Product Restrictions: (i) Customer shall not make products or any portion of them, in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier; and (ii) Customer is familiar with Supplier’s restrictions on the use of the Products and shall use the Products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').