Specifications
- Désignation:RoboSep™-C human CD8+ T cell Isolation kit, 1×10¹⁰ cells
- Cdt:1 KIT
Specifications
About this item
Immunomagnetic negative isolation of untouched CD8+ T cells from leukopaks.
- Fully automated large-scale isolation in a closed system
- Up to 94% purity with high recovery
- Untouched, viable cells
The RoboSep™-C Human CD8+ T Cell Isolation Kit is designed to isolate untouched and highly purified CD8+ T cells from fresh human peripheral blood leukopaks using RoboSep™-C (STMC100-0185 and #100-0186) for automated immunomagnetic negative selection.
This kit targets non-CD4+ T cells for removal with antibodies recognising specific cell surface markers. Using RoboSep™-C in a fully automated protocol, unwanted cells are labeled with antibodies and magnetic particles, and separated without columns using a RoboSep™-C magnet. The desired untouched cells are washed and concentrated by RoboSep™-C and transferred to a final product bag. Isolated cells are immediately available for downstream applications such as flow cytometry, culture, or cryopreservation.