Spécifications
- Désignation:EasySep™ extracellular vesicle PE positive selection kit, 1 kit
- Cdt:1 KIT
Spécifications
À propos de cet article
Fast and easy isolation of extracellular vesicles using immunomagnetic positive selection.
- Fast and easy-to-use
- No columns required
Efficiently obtain specific extracellular vesicle (EV) subtypes for use in downstream analyses or to enhance the sensitivity of your biomarker discovery assays. Use EasySep™ Extracellular Vesicle PE Positive Selection Kit to isolate tissue- and cell-specific EV subtypes from a variety of biofluids or cell-culture-conditioned medium in just 70 minutes.
EasySep™ Extracellular Vesicle PE Positive Selection Kit can be used with your choice of PE-conjugated antibodies to immunomagnetically isolate EV subtypes based on EV-associated surface markers. Compared to bulk EV isolation techniques (e.g. differential ultracentrifugation), the fast, easy, and column-free protocol results in higher enrichment of EV subtypes, making it easier to detect low frequency EVs in downstream analyses. Isolated EVs remain intact and are compatible with downstream analyses such as DNA/RNA extraction, western blot, or mass spectrometry.