MegaCult™-C Staining Kit for CFU-Mk
Catalog # STMC04962
Supplier: STEMCELL Technologies
CAS Number:
Specifications
- Cdt:1 KIT
- Désignation:MegaCult™-C Staining kit for CFU-Mk
Specifications
About this item
Staining kit for human CFU-Mk assays - 1 kit.
Stain fixed and dehydrated CFU-Mk colonies cultured in MegaCult™-C collagen-based medium using the reagents included in MegaCult™-C staining kit for CFU-Mk.
Megakaryocytes and platelets expressing the glycoprotein IIb/IIIa are stained with anti-human CD41 antibody and an alkaline phosphatase detection system. The kit is also suitable for the detection of bipotential erythroid-megakaryocyte progenitor cells (BFU-E/Mk).
Delivery: Kit includes MegaCult™-C Primary antibody, MegaCult™-C Control antibody, MegaCult™-C Human serum, MegaCult™-C Alkaline phosphatase substrate tablets, MegaCult™-C Biotin-conjugated goat anti-mouse IgG, MegaCult™-C Evans blue stain, MegaCult™-C 10% BSA.
Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').