Proteinase K Solution
STMC79016
:
- Cdt:4 mL
For removing nucleases from DNA/RNA samples and terminating enzymatic reactions.
Proteinase K is a serine protease that exhibits broad cleavage activity, cutting peptide bonds adjacent to the carboxylic group in aliphatic and aromatic amino acids. Proteinase K is used for a variety of applications, including removal of nucleases from DNA and RNA samples and terminating enzymatic reactions. Proteinase K Solution is in a ready-to-use format that is stable at room temperature and active over a pH range of 4,3 to 12,0 in 0,5% SDS or 1% Triton® X-100. Product is free of nuclease activity. Proteinase K Solution is required for use with ArciTect™ T7 Endonuclease I Kit.
: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').