ArciTect™ Annealing Buffer (5X)
ArciTect™ Annealing Buffer (5X)
ArciTect™ Annealing buffer (5X), 1 mL
N° de catalogue STMC76020
Marketsource
Fournisseur:   Stemcell Technologies
ArciTect™ Annealing Buffer (5X)
N° de catalogue STMC76020
Fournisseur:  Stemcell Technologies
Marketsource
Numéro CAS:  
ArciTect™ Annealing Buffer (5X)
ArciTect™ Annealing Buffer (5X)
ArciTect™ Annealing buffer (5X), 1 mL
Numéro CAS:  
Prix
21,00 €
Connexion Pour la disponibilité
1 * 1 mL
Product Details & Documents
For guide RNA generation in CRISPR-Cas9 genome editing.
ArciTect™ Annealing Buffer (5X) is required to anneal ArciTect™ tracrRNA and ArciTect™ crRNA to form a guide RNA (gRNA) duplex. This product is a refill component for ArciTect™ tracrRNA kit.
Attention: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Spécifications
Spécifications
  • Cdt:
    1 mL
  • Désignation:
    ArciTect™ Annealing buffer (5X)
  • Cat. No.:
    STMC76020
  • Spécificité cellulaire:
    Pluripotent stem cells