FluoroQuest™ TSA/PSA Antifade Mounting Medium *Optimised for Tyramide and Styramide Imaging*
N° de catalogue AATB44890
Fournisseur: AAT Bioquest
New Product
Numéro CAS:
Spécifications
- Cdt:5 mL
- Désignation:FluoroQuest™ TSA/PSA Antifade mounting medium *Optimised for Tyramide and Styramide imaging*
Spécifications
À propos de cet article
FluoroQuest™ TSA/PSA Antifade mounting medium is the only mounting medium that is optimised for tyramide (TSA) and styramide (PSA)-based fluorescence imaging applications.
- Stable for 12 months
Quality tested in-house.
Attention: For research use only.