Portelite™ Fluorimetric Lithium Ion Quantification Kit
Catalog # AATB21353
Supplier: AAT BIOQUEST
New Product
CAS Number:
Specifications
- Cdt:50 Tests
- Désignation:Portelite™ Fluorimetric Lithium Ion Quantification Kit
Specifications
About this item
Designed for consistent and dependable results. Portelite™ fluorimetric lithium ion quantification kit uses our robust lithium ion indicator dye, Lithiumighty™ 520, which exhibit great fluorescence intensity enhancement upon binding to lithium ions.
- Lithiumighty ™ 520 is perhaps the most robust lithium-ion indicator with high selectivity
- Rapid determination of lithium concentrations in a variety of samples
- 12-month shelf life
This nanodrop-based assay kit requires a small amount of sample, it is particularly suitable for the determination of lithium ion concentration in fields or on site.
Delivery: Shipped at RT.
Caution: Research use only.