ReadiUse™ NADP Regenerating Kit
Catalog # AATB15266
Supplier: AAT BIOQUEST
New Product
CAS Number:
Specifications
- Cdt:200 Tests
Specifications
About this item
Designed for consistent and dependable results. AAT Bioquest's ReadiUse™ NADP regenerating kit provides a flexible prepackaged NADP regenerating system to accommodate various experimental designs.
- Uses two ready-to-use solutions to regenerate NADP by a simple mixing
- 300-500 enzyme assays can be performed using this kit
- Six-month shelf life
This kit can be used for all NADP-requiring systems (such as: 12-Ketochenodeoxycholic Acid synthesis, Ferredoxin Reductase System).
Delivery: Shipped with blue ice.
Caution: Research use only.