Amplite® Fluorimetric Lithium Ion Quantification Kit
100 tests, 100 Tests
N° de catalogue AATB21351
Fournisseur: AAT Bioquest
Amplite® Fluorimetric Lithium Ion Quantification Kit
N° de catalogue AATB21351
Fournisseur: AAT Bioquest
Numéro CAS:
Aller à :
Product Details & Documents
Designed for consistent and dependable results. Amplite® fluorimetric lithium ion quantification kit uses our new robust lithium-ion indicator dye, Lithiumighty™ 520, which exhibits great fluorescence intensity enhancement upon binding to lithium ions.
- Lithiumighty ™ 520 is perhaps the most robust lithium-ion indicator with high selectivity
- Requires very small amount of sample
- 12 month shelf life
Microplate-based assay kit is particularly suitable for the determination of lithium ion concentration in a microvolume format.
Livraison: Shipped at RT.
Attention: Research use only.
Spécifications
Spécifications
- Cdt:100 Tests
- Tests par set:100 tests