ContactUsLinkComponent
Gelite™ 100 bp to 1 kb DNA Ladder
Gelite™ 100 bp to 1 kb DNA Ladder
N° de catalogue AATB17760
Fournisseur:  AAT BIOQUEST
New Product
Numéro CAS:  
undefined
Gelite™ 100 bp to 1 kb DNA Ladder
N° de catalogue AATB17760
Fournisseur:  AAT BIOQUEST
New Product
Numéro CAS:  
Spécifications
  • Cdt:
    0,5 mL
  • Désignation:
    Gelite™ 100 bp - 1 kb DNA ladder
  • Environmentally Preferable:
Spécifications
À propos de cet article
The Gelite™ 100 bp to 1 kb DNA ladder is designed to help size and estimate the mass of double-stranded DNA and PCR products within the 100 to 10000 base pair range on agarose or polyacrylamide gels.
  • Stable for 12 months
Quality tested in-house.
Attention: For research use only.