Anti-Persephin receptor Rabbit Polyclonal Antibody
Catalogus # ANTIA101081-100
Leverancier: ANTIBODIES.COM
New Product
Specificaties
- Type d'anticorps:Primaire
- Nom de l'antigène:GDNF family receptor alpha 4
- Symbole de l'antigène:Persephin receptor
- Clonalité:Polyclonal
- Conjugaison:Unconjugated
- ELISA:Yes
- Hôte:Rabbit
- Isotype:IgG
- Réactivité:Human
- Western blot:Yes
- Formulaire:Liquid
- ID de gène:UniprotID# Q9GZZ7
- Synonymes antigène:GFR-alpha-4|GFRA4_HUMAN|GFR alpha 4|GFR receptor alpha 4|GDNF family receptor alpha-4|GDNF receptor alpha-4|GDNFR-alpha-4|Glial cell line derived neurotrophic factor family receptor alpha 4|GFRA4
- Storage buffer:Supplied in Phosphate Buffered Saline (without Mg2+ and Ca2+), pH 7,4, with 150 mM NaCl, 0,02% Sodium Azide, and 50% Glycerol
- Molecular weight:31 kDa
- Température de stockage:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Température de transport:Shipped on blue ice at 4 °C
- Immunogène:Synthetic peptide derived from human GFRA4 (amino acids 141 - 190)
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunizing peptide
- Type de conditionnement:Plastic vial
- Cdt:100 µG
Specificaties
Over dit artikel
Rabbit polyclonal antibody to GFRA4 for WB and ELISA with samples derived from human.
- Validated Applications: WB, ELISA
- Recommended Dilutions: WB: 1:500 to 1:1000, ELISA: 1:10000
Type: Primary
Antigen: Persephin receptor
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human