Anti-Claudin 8 Rabbit Polyclonal Antibody
ANTIA101144-100
New Product
- Type d'anticorps:Primaire
- Nom de l'antigène:Claudin 8
- Symbole de l'antigène:Claudin 8
- Clonalité:Polyclonal
- Conjugaison:Unconjugated
- ELISA:Yes
- Hôte:Rabbit
- Isotype:IgG
- Réactivité:Human
- Western blot:Yes
- Environmentally Preferable:
- Formulaire:Liquid
- ID de gène:UniprotID# P56748
- Synonymes antigène:Cldn8|HEL S 79|OTTHUMP00000101915|Claudin-8|CLD8_HUMAN|Epididymis secretory protein Li 79
- Storage buffer:Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7,4, with 150 mM NaCl, 0,02% Sodium Azide, and 50% Glycerol
- Molecular weight:24 kDa
- Température de stockage:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Température de transport:Shipped on blue ice at 4 °C
- Immunogène:Synthetic peptide derived from human CLDN8 (amino acids 81-130)
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunizing peptide
- Type de conditionnement:Plastic vial
- Cdt:100 µG
Rabbit polyclonal antibody to CLDN8 for WB and ELISA with samples derived from human.
- Validated Applications: WB, ELISA
- Recommended Dilutions: WB: 1:500 to 1:1000, ELISA: 1:1000
Type: Primary
Antigen: Claudin 8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human