Order Entry
Belgium
ContactUsLinkComponent
Anti-Arginase 1 Rabbit Recombinant Antibody [clone: ARG1/6790R]
Anti-Arginase 1 Rabbit Recombinant Antibody [clone: ARG1/6790R]
N° de catalogue ANTIA278012-100
Fournisseur:  ANTIBODIES.COM
New Product
undefined
Anti-Arginase 1 Rabbit Recombinant Antibody [clone: ARG1/6790R]
N° de catalogue ANTIA278012-100
Fournisseur:  ANTIBODIES.COM
New Product

Spécifications

  • Type d'anticorps:
    Primaire
  • Nom de l'antigène:
    A I
  • Symbole de l'antigène:
    Arginase 1
  • Clonalité:
    Recombinant
  • Clone:
    ARG1/6790R
  • Conjugaison:
    Unconjugated
  • Hôte:
    Rabbit
  • ImmunoChimie:
    Yes
  • Isotype:
    IgG
  • Réactivité:
    Human
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Formulaire:
    Liquid
  • ID de gène:
    UniprotID# P05089
  • Synonymes antigène:
    Al|ARG 1|Arginase-1|Arginase 1|Arginase type I|ARGI1_HUMAN|Arginase, liver|Arginase liver|arg1
  • Storage buffer:
    Supplied in 10 mM phosphate buffered saline with 0,05% BSA and 0,05% sodium azide
  • Molecular weight:
    35 - 38 kDa
  • Température de stockage:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Concentration:
    200 µg/ml
  • Température de transport:
    Shipped on blue ice at 4 °C
  • Immunogène:
    Recombinant fragment, around amino acids 300-400, of human Arginase 1. The exact sequence is proprietary.
  • Tested applications:
    IHC
  • Purification:
    Protein A/G chromatography
  • Type de conditionnement:
    Plastic vial
  • Cdt:
    100 µG

Spécifications

À propos de cet article

Recombinant rabbit monoclonal [ARG1/6790R] antibody to Arginase 1 for WB and IHC with samples derived from human.

  • Validated Applications: WB, IHC
  • Recommended Dilutions: WB: 2 to 4 µg/ml, IHC-P: 2 to 4 µg/ml

Type: Primary
Antigen: Arginase 1
Clonality: Recombinant
Clone: ARG1/6790R
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human