Anti-His Tag Rabbit Recombinant Antibody [clone: DM1]
New Product
Recombinant rabbit monoclonal [DM1] antibody to His Tag for ELISA and WB.
- Validated Applications: ELISA, WB
- Recommended Dilutions: WB: 1:1,000
- Host: Rabbit
- Clonality: Monoclonal
- Isotype: IgG
- Conjugate: Unconjugated
Type: Primary
Antigen: His Tag
Clonality: Recombinant
Clone: DM1
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
- Catalog No:
- ANTIA317439-500
- ANTIA317439-100
- Symbole de l'antigène:
- His Tag
- His Tag
- Nom de l'antigène:
- 6 His epitope tag
- 6 His epitope tag
- Synonymes antigène:
- HHHHHH tag|Polyhistidine Tag|HHHHHH epitope tag|Hexa His tag|6X His tag
- HHHHHH tag|Polyhistidine Tag|HHHHHH epitope tag|Hexa His tag|6X His tag
- Type d'anticorps:
- Primaire
- Primaire
- Clonalité:
- Recombinant
- Recombinant
- Conjugaison:
- Unconjugated
- Unconjugated
- Clone:
- DM1
- DM1
- Hôte:
- Rabbit
- Rabbit
- Isotype:
- IgG
- IgG
- Western blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- Formulaire:
- Lyophilised
- Lyophilised
- Purification:
- Affinity chromatography
- Affinity chromatography
- Concentration:
- Reconstitution dependent
- Reconstitution dependent
- Storage buffer:
- Lyophilised from sterile phosphate buffered saline, pH 7,4. Normally 5 - 8% Trehalose is added as a protectant before lyophilisation
- Lyophilised from sterile phosphate buffered saline, pH 7,4. Normally 5 - 8% Trehalose is added as a protectant before lyophilisation
- Température de stockage:
- Shipped at 4 °C. Lyophilised: Store at –20...–80 °C. Reconstituted: Aliquot and store at –80 °C. Product is stable for one year. Avoid freeze/thaw cycles.
- Shipped at 4 °C. Lyophilised: Store at –20...–80 °C. Reconstituted: Aliquot and store at –80 °C. Product is stable for one year. Avoid freeze/thaw cycles.
- Température de transport:
- Shipped on blue ice at 4 °C
- Shipped on blue ice at 4 °C
Frequently Bought Together