Anti-Biotin Mouse Monoclonal Antibody [Clone: BTN/403]
Catalog # ANTIA277867-100
Supplier: ANTIBODIES.COM
New Product
Specifications
- Type d'anticorps:Primaire
- Nom de l'antigène:Biotin
- Symbole de l'antigène:Biotin
- Clonalité:Monoclonal
- Clone:BTN/403
- Conjugaison:Unconjugated
- Cytométrie en flux:Yes
- Hôte:Mouse
- ImmunoChimie:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG1 kappa
- Réactivité:
- Western blot:Yes
- Environmentally Preferable:
- Formulaire:Liquid
- Synonymes antigène:Biotin
- Storage buffer:Supplied in 10 mM phosphate buffered saline with 0,05% BSA and 0,05% sodium azide
- Molecular weight:244Da
- Température de stockage:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:200 µg/ml
- Température de transport:Shipped on blue ice at 4 °C
- Immunogène:Biotinylated sheep immunoglobulin.
- Tested applications:IHC-P
- Purification:Protein A/G chromatography
- Type de conditionnement:Plastic vial
- Cdt:100 µG
Specifications
About this item
Mouse monoclonal [BTN/403] antibody to Biotin for Flow Cytometry, IF, WB and IHC-P.
- Validated Applications: Flow Cytometry, IF, WB, IHC-P
- Recommended Dilutions: Flow Cytometry: 0,5 to 1 µg/million cells, IF: 0,5 to 1 µg/ml, WB: 0,5 to 2 µg/ml, IHC-P: 0,5 to 1 µg/ml
Type: Primary
Antigen: Biotin
Clonality: Monoclonal
Clone: BTN/403
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: