Anti-CD28 Mouse Monoclonal Antibody [Clone: C28/75]
N° de catalogue ANTIA253810-100
Fournisseur: ANTIBODIES.COM
New Product
Spécifications
- Type d'anticorps:Primaire
- Nom de l'antigène:CD 28
- Symbole de l'antigène:CD28
- Clonalité:Monoclonal
- Clone:C28/75
- Conjugaison:Unconjugated
- Cytométrie en flux:Yes
- Hôte:Mouse
- Isotype:IgG1 kappa
- Réactivité:Human
- Environmentally Preferable:
- Formulaire:Liquid
- ID de gène:UniprotID# P10747
- Synonymes antigène:CD28|T cell specific surface glycoprotein CD28|MGC138290|T-cell-specific surface glycoprotein CD28|CD28_HUMAN|T cell specific surface glycoprotein|CD28 molecule|CD28 antigen|T cell antigen CD28
- Storage buffer:Supplied in 10 mM phosphate buffered saline; without sodium azide and carrier free
- Molecular weight:44 kDa (monomer)/88 kDa (dimer)
- Température de stockage:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:1 mg/ml
- Température de transport:Shipped on blue ice at 4 °C
- Immunogène:Recombinant human CD28 protein.
- Tested applications:Functional studies
- Purification:Protein A/G chromatography
- Type de conditionnement:Plastic vial
- Cdt:100 µG
Spécifications
À propos de cet article
Mouse monoclonal [C28/75] antibody to CD28 for Flow Cytometry and Functional Studies with samples derived from human.
- Validated Applications: Flow Cytometry, Functional Studies
- Recommended Dilutions: Flow Cytometry: 1 to 2 µg/million cells
Type: Primary
Antigen: CD28
Clonality: Monoclonal
Clone: C28/75
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human