Order Entry
Belgium
ContactUsLinkComponent
 
Anti-14-3-3 gamma/YWHAG Rabbit Monoclonal Antibody [clone: ARC1457]
  ANTIA309079-100
New Product
undefined
Anti-14-3-3 gamma/YWHAG Rabbit Monoclonal Antibody [clone: ARC1457]
  ANTIA309079-100
New Product

 

  • Type d'anticorps:
    Primaire
  • Nom de l'antigène:
    14-3-3 gamma / YWHAG
  • Symbole de l'antigène:
    14-3-3 gamma / YWHAG
  • Clonalité:
    Monoclonal
  • Clone:
    ARC1457
  • Conjugaison:
    Unconjugated
  • Hôte:
    Rabbit
  • ImmunoChimie:
    Yes
  • Isotype:
    IgG
  • Réactivité:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Formulaire:
    Liquid
  • ID de gène:
    UniprotID# P61981
  • Storage buffer:
    Supplied in phosphate buffered saline, pH 7,3, with 50% glycerol, 0,05% BSA, and 0,02% sodium azide
  • Molecular weight:
    28 kDa
  • Sequence:
    VLSLLDNYLIKNCSETQYESKVFYLKMKGDYYRYLAEVATGEKRATVVESSEKAYSEAHEISKEHMQPTHPIRLGLALNYSVFYYEIQNAPEQACHLAKTA
  • Température de stockage:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Température de transport:
    Shipped on blue ice at 4 °C
  • Immunogène:
    A synthetic peptide corresponding to a sequence within amino acids 100-200 of human 14-3-3 gamma (P61981).
  • Tested applications:
    IHC
  • Purification:
    Affinity purification
  • Type de conditionnement:
    Plastic vial
  • Cdt:
    100 µl

 

 

Rabbit monoclonal [ARC1457] antibody to 14-3-3 gamma/YWHAG for WB and IHC with samples derived from human, mouse and rat.

  • Validated applications: WB, IHC
  • Recommended dilutions: WB: 1:500 to 1:2,000, IHC: 1:50 to 1:200

Type: Primary
Antigen: 14-3-3 gamma / YWHAG
Clonality: Monoclonal
Clone: ARC1457
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat