Anti-BRG1 Rabbit Recombinant Antibody [clone: BRG1/7633R]
ANTIA314527-100
New Product
- Type d'anticorps:Primaire
- Nom de l'antigène:ATP dependent helicase SMARCA4
- Symbole de l'antigène:BRG1
- Clonalité:Recombinant
- Clone:BRG1/7633R
- Conjugaison:Unconjugated
- ELISA:Yes
- Hôte:Rabbit
- ImmunoChimie:Yes
- Isotype:IgG kappa
- Réactivité:Human
- Western blot:Yes
- Formulaire:Liquid
- ID de gène:UniprotID# P51532
- Synonymes antigène:BAF 190|Brahma protein like 1|ATP-dependent helicase SMARCA4|BRG1 protein|BAF190|BRG1|BAF190A|BRG1-associated factor 190A|BRG1 associated factor 190A
- Storage buffer:Supplied in 10 mM phosphate buffered saline with 0,05% BSA and 0,05% sodium azide
- Molecular weight:185 kDa
- Température de stockage:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:200 µg/ml
- Température de transport:Shipped on blue ice at 4 °C
- Immunogène:Recombinant fragment (around Amino Acids 200-400) of human SMARCA4 protein. The exact sequence is proprietary.
- Tested applications:IHC-P
- Purification:Protein A/G chromatography
- Type de conditionnement:Plastic vial
- Cdt:100 µG
Recombinant rabbit monoclonal [BRG1/7633R] antibody to BRG1 for ELISA, WB and IHC-P with samples derived from human.
- Validated applications: ELISA, WB, IHC-P
- Recommended dilutions: WB: 1 to 2 µg/ml, IHC: 1 to 2 µg/ml
Type: Primary
Antigen: BRG1
Clonality: Recombinant
Clone: BRG1/7633R
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG kappa
Reactivity: Human