Order Entry
Belgium
ContactUsLinkComponent
 
Anti-RAB11-FIP2 Rabbit Polyclonal Antibody
  ANTIA97814-100
New Product
undefined
Anti-RAB11-FIP2 Rabbit Polyclonal Antibody
  ANTIA97814-100
New Product

 

  • Type d'anticorps:
    Primaire
  • Nom de l'antigène:
    KIAA0941
  • Symbole de l'antigène:
    RAB11-FIP2
  • Clonalité:
    Polyclonal
  • Conjugaison:
    Unconjugated
  • ELISA:
    Yes
  • Hôte:
    Rabbit
  • Isotype:
    IgG
  • Réactivité:
    Human,
    Mouse
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Formulaire:
    Liquid
  • ID de gène:
    UniprotID# Q7L804
  • Synonymes antigène:
    RFIP2_HUMAN|RAB11-FIP2|RFIP2|NRip11|RAB11 family interacting protein 2 (class I)|RAB11FIP2|RAB11 FIP2 long isoform|Rab11 family-interacting protein 2
  • Storage buffer:
    Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7,4, with 150 mM NaCl, 0,02% Sodium Azide, and 50% Glycerol
  • Molecular weight:
    58 kDa
  • Température de stockage:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Température de transport:
    Shipped on blue ice at +4 °C
  • Immunogène:
    Synthetic peptide derived from human RAB11FIP2 (amino acids 340-389)
  • Purification:
    Purified from rabbit serum by antigen affinity chromatography using the immunising peptide
  • Type de conditionnement:
    Plastic vial
  • Cdt:
    100 µG

 

 

Rabbit polyclonal antibody to RAB11FIP2 for WB and ELISA with samples derived from Human and Mouse.

  • Validated Applications: WB, ELISA
  • Recommended Dilutions: WB: 1:500 to 1:1000, ELISA: 1:1000

Type: Primary
Antigen: RAB11-FIP2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse