Saisie de commande
Belgium
Lien pour commandesContactUsLinkComponent
Anti-S100A7L2 Rabbit Polyclonal Antibody

Anti-S100A7L2 Rabbit Polyclonal Antibody

Plastic vial
N° de catalogue ANTIA99949-100
Fournisseur:   Antibodies.com
Anti-S100A7L2 Rabbit Polyclonal Antibody
N° de catalogue ANTIA99949-100
Fournisseur:  Antibodies.com
Anti-S100A7L2 Rabbit Polyclonal Antibody
Anti-S100A7L2 Rabbit Polyclonal Antibody

Plastic vial

Prix
495,00 €
Connexion Pour la disponibilité
1 * 1 EA
Product Details & Documents

Rabbit polyclonal antibody to S100A7L2 for WB and ELISA with samples derived from Human.

  • Recommended dilutions: WB: 1:500 to 1:1000; ELISA: 1:10000

Attention: This product is for research use only. It is not for use in diagnostic or therapeutic procedures.

Type: Primary
Antigen: S100A7L2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human

Spécifications
  • Seq:
    1286
  • Type de conditionnement:
    Plastic vial
  • Western blot:
    Yes
  • Type d'anticorps:
    Primaire
  • Formulaire:
    Liquid
  • Purification:
    Purified from rabbit serum by antigen affinity chromatography using the immunising peptide
  • Symbole de l'antigène:
    S100A7L2
  • Synonymes antigène:
    S100 calcium binding protein A7 like 2|S100A7B
  • Isotype:
    IgG
  • Masse moléculaire:
    11 kDa
  • Conjugaison:
    Unconjugated
  • Réactivité:
    Human
  • Température de stockage:
    Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • ID de gène:
    UniprotID# Q5SY68
  • Cat. No.:
    ANTIA99949-100
  • Clonalité:
    Polyclonal
  • Immunogène:
    Synthetic peptide derived from human S100A7L2 (amino acids 38-87)
  • Nom de l'antigène:
    Protein S100 A7 like 2
  • Hôte:
    Rabbit
  • Storage buffer:
    Supplied in phosphate buffered saline (without Mg2+ and Ca2+), pH 7,4, with 150 mM NaCl, 0,02% sodium azide, and 50% glycerol
  • Température de transport:
    Shipped on blue ice at 4 °C
  • ELISA:
    Yes