Order Entry
Belgium
ContactUsLinkComponent
Anti-TMF Rabbit Polyclonal Antibody
Anti-TMF Rabbit Polyclonal Antibody
  ANTIA307488-100
New Product
undefined
Anti-TMF Rabbit Polyclonal Antibody
  ANTIA307488-100
New Product

 

  • Type d'anticorps:
    Primaire
  • Symbole de l'antigène:
    TMF
  • Clonalité:
    Polyclonal
  • Conjugaison:
    Unconjugated
  • Hôte:
    Rabbit
  • Isotype:
    IgG
  • Réactivité:
    Human,
    Mouse
  • Western blot:
    Yes
  • Formulaire:
    Liquid
  • ID de gène:
    UniprotID# P82094
  • Storage buffer:
    Supplied in Phosphate buffered saline, pH 7,3, with 50% glycerol and 0,01% Thiomersal
  • Molecular weight:
    160 kDa
  • Sequence:
    MKVEQERKKAIFTQETIKEKERKPFSVSSTPTMSRSSSISGVDMAGLQTSFLSQDESHDHSFGPMPISANGSNLYDAVRMGAGSSIIENLQSQLKLREGEI
  • Température de stockage:
    Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze/thaw cycles.
  • Température de transport:
    Shipped on blue ice at 4 °C
  • Immunogène:
    A synthetic peptide corresponding to a sequence within amino acids 900-1000 of human TMF1 (NP_009045.2).
  • Purification:
    Affinity purification
  • Type de conditionnement:
    Plastic vial
  • Cdt:
    100 µl

 

 

Rabbit polyclonal antibody to TMF for WB with samples derived from Human and Mouse.

  • Validated Applications: WB
  • Recommended Dilutions: WB: 1:500 to 1:2000

Type: Primary
Antigen: TMF
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse