Order Entry
Belgium
ContactUsLinkComponent
Anti-GITR Rabbit Recombinant Antibody [clone: DM79] (Biotinylated)
Anti-GITR Rabbit Recombinant Antibody [clone: DM79] (Biotinylated)
Supplier:  ANTIBODIES.COM
New Product
Certificates
Anti-GITR Rabbit Recombinant Antibody [clone: DM79] (Biotinylated)
Supplier:  ANTIBODIES.COM
New Product

Specifications

  • Catalog No:
  • ANTIA317380-500
  • ANTIA317380-100
  • Symbole de l'antigène:
  • GITR
  • GITR
  • Nom de l'antigène:
  • Activation inducible TNFR family receptor
  • Activation inducible TNFR family receptor
  • Synonymes antigène:
  • CD357|GITR|GITRD|Activation-inducible TNFR family receptor|Glucocorticoid induced TNFR related protein|GITR D|AITR|GITR-D|glucocorticoid-induced tnf receptor ligand
  • CD357|GITR|GITRD|Activation-inducible TNFR family receptor|Glucocorticoid induced TNFR related protein|GITR D|AITR|GITR-D|glucocorticoid-induced tnf receptor ligand
  • Type d'anticorps:
  • Primaire
  • Primaire
  • Clonalité:
  • Recombinant
  • Recombinant
  • Conjugaison:
  • Biotin
  • Biotin
  • Clone:
  • DM79
  • DM79
  • Réactivité:
  • Human
  • Human
  • Hôte:
  • Rabbit
  • Rabbit
  • ID de gène:
  • UniprotID# Q9Y5U5
  • UniprotID# Q9Y5U5
  • Isotype:
  • IgG
  • IgG
  • ELISA:
  • Yes
  • Yes
  • Cytométrie en flux:
  • Yes
  • Yes
  • Formulaire:
  • Lyophilised
  • Lyophilised
  • Purification:
  • Affinity chromatography
  • Affinity chromatography
  • Concentration:
  • Reconstitution dependent
  • Reconstitution dependent
  • Storage buffer:
  • Lyophilised from sterile Phosphate Buffered Saline, pH 7,4. Normally 5 - 8% Trehalose is added as a protectant before lyophilisation
  • Lyophilised from sterile Phosphate Buffered Saline, pH 7,4. Normally 5 - 8% Trehalose is added as a protectant before lyophilisation
  • Température de stockage:
  • Shipped at 4 °C. Lyophilised: Store at –20...–80 °C. Reconstituted: Aliquot and store at –80 °C. Product is stable for one year. Avoid freeze / thaw cycles.
  • Shipped at 4 °C. Lyophilised: Store at –20...–80 °C. Reconstituted: Aliquot and store at –80 °C. Product is stable for one year. Avoid freeze / thaw cycles.
  • Température de transport:
  • Shipped on blue ice at 4 °C
  • Shipped on blue ice at 4 °C

Specifications

Frequently Bought Together