Anti-PCNA Mouse Recombinant Antibody [clone: rPCNA/8858]
ANTIA316866-100
New Product
- Type d'anticorps:Primaire
- Nom de l'antigène:ATLD2
- Symbole de l'antigène:PCNA
- Clonalité:Recombinant
- Clone:rPCNA/8858
- Conjugaison:Unconjugated
- Hôte:Mouse
- ImmunoChimie:Yes
- Isotype:IgG1 kappa
- Réactivité:Human
- Formulaire:Liquid
- ID de gène:UniprotID# P12004
- Synonymes antigène:DNA polymerase delta auxiliary protein|Mutagen-sensitive 209 protein|etID36690.10|HGCN8729|MGC8367|cb16|fa28e03|fb36g03|Cyclin
- Storage buffer:Supplied in 10 mM phosphate buffered saline; without sodium azide and carrier free.
- Molecular weight:36 kDa
- Température de stockage:Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze / thaw cycles.
- Concentration:1 mg/ml
- Température de transport:Shipped on blue ice at +4 °C
- Immunogène:Recombinant full-length human PCNA protein.
- Tested applications:IHC-P
- Purification:Protein A/G chromatography
- Type de conditionnement:Plastic vial
- Cdt:100 µG
Recombinant mouse monoclonal [rPCNA/8858] antibody to PCNA for IHC-P with samples derived from human.
- Validated applications: IHC-P
- Recommended dilutions: IHC: 1 to 2 µg/ ml
Type: Primary
Antigen: PCNA
Clonality: Recombinant
Clone: rPCNA/8858
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human