Order Entry
Belgium
ContactUsLinkComponent
Anti-Inhibin alpha Rabbit Recombinant Antibody [clone: INHA/8872R]
Anti-Inhibin alpha Rabbit Recombinant Antibody [clone: INHA/8872R]
  ANTIA316569-100
New Product
Anti-Inhibin alpha Rabbit Recombinant Antibody [clone: INHA/8872R]
  ANTIA316569-100
New Product

 

  • Type d'anticorps:
    Primaire
  • Nom de l'antigène:
    A inhibin subunit
  • Symbole de l'antigène:
    Inhibin alpha
  • Clonalité:
    Recombinant
  • Clone:
    INHA/8872R
  • Conjugaison:
    Unconjugated
  • Hôte:
    Rabbit
  • ImmunoChimie:
    Yes
  • Isotype:
    IgG kappa
  • Réactivité:
    Human
  • Environmentally Preferable:
  • Formulaire:
    Liquid
  • ID de gène:
    UniprotID# P05111
  • Synonymes antigène:
    A inhibin subunit precursor|Inhibin alpha subunit|INHA_HUMAN|IHA|Inhibin alpha chain precursor|inhA|Inhibin alpha chain|AW55578|Inhibin alpha
  • Storage buffer:
    Supplied in 10 mM phosphate buffered saline; without sodium azide and carrier free.
  • Molecular weight:
    47 kDa
  • Température de stockage:
    Shipped at 4 °C. Upon delivery aliquot and store at –20 °C. Avoid freeze / thaw cycles.
  • Concentration:
    1 mg/ml
  • Température de transport:
    Shipped on blue ice at +4 °C
  • Immunogène:
    Recombinant fragment of human Inhibin alpha protein (around Amino Acids 233-362). The exact sequence is proprietary.
  • Tested applications:
    IHC-P
  • Purification:
    Protein A/G chromatography
  • Type de conditionnement:
    Plastic vial
  • Cdt:
    100 µG

 

 

Recombinant rabbit monoclonal [INHA/8872R] antibody to Inhibin alpha for IHC-P with samples derived from human.

  • Validated applications: IHC-P
  • Recommended dilutions: IHC: 1 to 2 µg/ ml

Type: Primary
Antigen: Inhibin alpha
Clonality: Recombinant
Clone: INHA/8872R
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG kappa
Reactivity: Human