Human CRLF2 ELISA Kit
Human CRLF2 ELISA kit is a 90 minutes sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human CRLF2 in serum, plasma, and other biological fluids.
- Higher throughput: Get results in just 90 minutess, with a single wash step
- Detection range: 0,75 to 12 ng/ml
- Sensitivity: 0,026 ng/ml
- Sample Type: Serum, plasma or other biological fluids
- Assay Precision: Intra-assay: CV <8%, Inter-assay: CV <10%
Human CRLF2 ELISA kit employs the sandwich enzyme immunoassay technique for the quantitative measurement of human CRLF2 in serum, plasma or other biological fluids. An antibody specific for CRLF2 has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the CRLF2 present in each sample is bound to the wells by the immobilised antibody. Biotinylated anti-CRLF2 antibody, which also binds the CRLF2 present in each sample, and Streptavidin-HRP, which binds the Biotinylated anti-CRLF2 antibody, are added and the microtiter plate is incubated. Following incubation, unbound Biotinylated anti-CRLF2 antibody and unbound Streptavidin-HRP are removed by washing, and two substrate solutions are added to the wells. Colour develops in proportion to the amount of CRLF2 captured in each well. The colour development is stopped by addition of stop solution which changes the colour from blue to yellow and the intensity of the colour is then measured. The concentration of CRLF2 in the samples can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.
: This is a ready-to-use ELISA kit that contains: Pre-coated 96 well microplate, standard solution, standard diluent, biotinylated detection antibody, steptavidin-HRP, wash buffer (25X), substrate solution A, substrate solution B, stop solution, plate sealers, foil pouch, and protocol manual.
- Cdt:96 Tests
- Désignation:Human CRLF2 ELISA kit
- Type de test:Sandwich
- Méthode de détection:Colorimetric
- Primary antibody reactivity:Human
- Portée de détection:0,75 - 12 ng/ml
- Type déchantillon:Serum, plasma or other biological fluids
- Time to Results:1 h 30 min
- Température de stockage:4 °C
- Cat. No.:ANTIA312760-96
- Volume d'échantillon:40 μl
- Format:Pre-coated
- Cible:CRLF2
- Sensibilité:0,026 ng/ml
- Durée du test:One step
- Regulatory status:RUO
- Durée de conservation:Store for 6 months at 4 °C
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



