Mouse Rheumatoid Factor IgM ELISA Kit
Catalog # ANTIA247075-96
Supplier: ANTIBODIES.COM
New Product
CAS Number:
Specifications
- Cdt:96 T
- Durée du test:Multiple steps
- Type de test:Indirect
- Format:Pre-coated
- Primary antibody reactivity:Mouse
- Cible:Rheumatoid Factor IgM
- Désignation:Mouse rheumatoid factor IgM ELISA kit
- Type déchantillon:Serum, plasma, tissue homogenates, and other biological fluids
- Méthode de détection:Colorimetric
- Time to Results:4 h 30 min
- Durée de conservation:Store for 6 months at 4 °C
- Portée de détection:4,688 - 300 IU/ml
- Température de stockage:4 °C
- Sensibilité:2,813 IU/ml
- Regulatory status:RUO
Specifications
About this item
Mouse Rheumatoid Factor IgM ELISA kit is a indirect Enzyme-Linked Immunosorbent Assay designed for the in vitro quantitative determination of mouse Rheumatoid Factor IgM in serum, plasma, tissue homogenates, and other biological fluids.