Order Entry
Belgium
ContactUsLinkComponent
Monkey IL-1 Beta ELISA Kit
Monkey IL-1 Beta ELISA Kit
Catalog # ANTIA246983-96
Supplier:  ANTIBODIES.COM
New Product
CAS Number:  
undefined
Monkey IL-1 Beta ELISA Kit
Catalog # ANTIA246983-96
Supplier:  ANTIBODIES.COM
New Product
CAS Number:  

Specifications

  • Cdt:
    96 T
  • Durée du test:
    Multiple steps
  • Type de test:
    Sandwich
  • Format:
    Pre-coated
  • Primary antibody reactivity:
    Monkey
  • Cible:
    IL-1 beta
  • Désignation:
    Monkey IL-1 beta ELISA kit
  • Type déchantillon:
    Serum, plasma, tissue homogenates, and other biological fluids
  • Réactivité croisée:
    Monkey IL-1 beta ELISA Kit exhibits high specificity and excellent specificity for the detection of monkey IL-1 beta. No significant cross-reactivity or interference between IL-1 beta and analogues was observed.
  • Méthode de détection:
    Colorimetric
  • Time to Results:
    4 h 30 min
  • Durée de conservation:
    Store for 6 months at 4 °C
  • Portée de détection:
    3,906 - 250 pg/ml
  • Température de stockage:
    4 °C
  • Volume d'échantillon:
    100 μl
  • Sensibilité:
    2,344 pg/ml
  • Regulatory status:
    RUO

Specifications

About this item

Monkey IL-1 beta ELISA kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of monkey IL-1 beta in serum, plasma, tissue homogenates, and other biological fluids.

  • Ready-to-use ELISA kit
  • Assay precision: Intra-assay: CV <8%, Inter-assay: CV <10%

Monkey IL-1 beta ELISA kit employs the sandwich enzyme immunoassay technique for the quantitative measurement of monkey IL-1 beta in serum, plasma, tissue homogenates, and other biological fluids. An antibody specific for IL-1 beta has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the IL-1 beta present in each sample is bound to the wells by the immobilised antibody. Following incubation, the wells are washed and then incubated with Biotinylated Anti-IL-1 beta Antibody, which binds the captured IL-1 beta present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualise the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of IL-1 beta captured in each well. The concentration of IL-1 beta can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.

Ordering information: This is a ready-to-use ELISA kit that contains: Pre-coated 96 well microplate, lyophilised standard, sample dilution buffer, biotinylated detection antibody (100X), antibody dilution buffer, HRP-Streptavidin conjugate (SABC) (100X), SABC dilution buffer, TMB substrate, stop solution, wash buffer (25X), plate sealers, foil pouch, and protocol manual.