Spécifications
- Cdt:96 Tests
- Durée du test:One step
- Type de test:Sandwich
- Format:Pre-coated
- Hôte:
- Primary antibody reactivity:Human
- Cible:KDM3A/JHDM2A
- Désignation:Human KDM3A/JHDM2A ELISA kit
- Environmentally Preferable:
- Type déchantillon:Serum, plasma or other biological fluids
- Méthode de détection:Colorimetric
- Time to Results:1 h 30 min
- Durée de conservation:Store for 6 months at 4 °C
- Portée de détection:50 - 800 ng/L
- Température de stockage:4 °C
- Volume d'échantillon:40 μl
- Sensibilité:3,48 ng/L
- Regulatory status:RUO
Spécifications
À propos de cet article
Human KDM3A/JHDM2A ELISA kit is a 90 minutes sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human KDM3A/JHDM2A in serum, plasma, and other biological fluids.
- Higher throughput: Get results in just 90 minutess, with a single wash step
- Detection Range: 50 to 800 ng/L
- Sensitivity: 3,48 ng/L
- Sample Type: Serum, plasma or other biological fluids
- Assay Precision: Intra-assay: CV <8%, Inter-assay: CV <10%
Human KDM3A/JHDM2A ELISA kit employs the sandwich enzyme immunoassay technique for the quantitative measurement of human KDM3A/JHDM2A in serum, plasma or other biological fluids. An antibody specific for KDM3A/JHDM2A has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the KDM3A/JHDM2A present in each sample is bound to the wells by the immobilised antibody. Biotinylated anti-KDM3A/JHDM2A antibody, which also binds the KDM3A/JHDM2A present in each sample, and Streptavidin-HRP, which binds the Biotinylated anti-KDM3A/JHDM2A antibody, are added and the microtiter plate is incubated. Following incubation, unbound Biotinylated anti-KDM3A/JHDM2A antibody and unbound Streptavidin-HRP are removed by washing, and two substrate solutions are added to the wells. Colour develops in proportion to the amount of KDM3A/JHDM2A captured in each well. The colour development is stopped by addition of stop solution which changes the colour from blue to yellow and the intensity of the colour is then measured. The concentration of KDM3A/JHDM2A in the samples can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.