Order Entry
Belgium
ContactUsLinkComponent
Human IL-2 ELISA Kit
Human IL-2 ELISA Kit
Catalog # ANTIA310849-96
Supplier:  ANTIBODIES.COM
New Product
CAS Number:  
undefined
Human IL-2 ELISA Kit
Catalog # ANTIA310849-96
Supplier:  ANTIBODIES.COM
New Product
CAS Number:  

Specifications

  • Cdt:
    96 Tests
  • Durée du test:
    One step
  • Type de test:
    Sandwich
  • Format:
    Pre-coated
  • Hôte:
  • Primary antibody reactivity:
    Human
  • Cible:
    IL-2
  • Désignation:
    Human IL-2 ELISA kit
  • Environmentally Preferable:
  • Type déchantillon:
    Serum, plasma or other biological fluids
  • Méthode de détection:
    Colorimetric
  • Time to Results:
    1 h 30 min
  • Durée de conservation:
    Store for 6 months at 4 °C
  • Portée de détection:
    75 - 1200 ng/L
  • Température de stockage:
    4 °C
  • Volume d'échantillon:
    40 μl
  • Sensibilité:
    2,51 ng/L
  • Regulatory status:
    RUO

Specifications

About this item

Human IL-2 ELISA kit is a 90 minutes sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human IL-2 in serum, plasma, and other biological fluids.

  • Higher throughput: Get results in just 90 minutess, with a single wash step
  • Detection Range: 75 to 1200 ng/L
  • Sensitivity: 2,51 ng/L
  • Sample Type: Serum, plasma or other biological fluids
  • Assay Precision: Intra-assay: CV <8%, Inter-assay: CV <10%

Human IL-2 ELISA kit employs the sandwich enzyme immunoassay technique for the quantitative measurement of human IL-2 in serum, plasma or other biological fluids. An antibody specific for IL-2 has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the IL-2 present in each sample is bound to the wells by the immobilised antibody. Biotinylated anti-IL-2 antibody, which also binds the IL-2 present in each sample, and Streptavidin-HRP, which binds the Biotinylated anti-IL-2 antibody, are added and the microtiter plate is incubated. Following incubation, unbound Biotinylated anti-IL-2 antibody and unbound Streptavidin-HRP are removed by washing, and two substrate solutions are added to the wells. Colour develops in proportion to the amount of IL-2 captured in each well. The colour development is stopped by addition of stop solution which changes the colour from blue to yellow and the intensity of the colour is then measured. The concentration of IL-2 in the samples can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.

Ordering information: This is a ready-to-use ELISA kit that contains: Pre-coated 96 well microplate, standard solution, standard diluent, biotinylated detection antibody, streptavidin-HRP, wash buffer (25X), substrate solution A, substrate solution B, stop solution, plate sealers, foil pouch, and protocol manual.