Saisie de commande
Belgium
Lien pour commandesContactUsLinkComponent
 

Human CRBN ELISA Kit

Human CRBN ELISA kit, 96 Tests
Catalog # ANTIA311905-96
Supplier:   Antibodies.com
Human CRBN ELISA Kit
Catalog # ANTIA311905-96
Supplier:  Antibodies.com
CAS Number:  
Human CRBN ELISA Kit
Human CRBN ELISA Kit

Human CRBN ELISA kit, 96 Tests

CAS Number:  
Price
565,00 €
Log in for availability
1 * 96 Tests
Product Details & Documents

Human CRBN ELISA kit is a 90 minutes sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human CRBN in serum, plasma, and other biological fluids.

  • Higher throughput: Get results in just 90 minutess, with a single wash step
  • Detection range: 1,5 to 24 ng/ml
  • Sensitivity: 0,043 ng/ml
  • Sample Type: Serum, plasma or other biological fluids
  • Assay precision: Intra assay: CV <8%, Inter assay: CV <10%

Human CRBN ELISA kit employs the sandwich enzyme immunoassay technique for the quantitative measurement of human CRBN in serum, plasma or other biological fluids. An antibody specific for CRBN has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the CRBN present in each sample is bound to the wells by the immobilised antibody. Biotinylated anti-CRBN antibody, which also binds the CRBN present in each sample, and Streptavidin-HRP, which binds the Biotinylated anti-CRBN antibody, are added and the microtiter plate is incubated. Following incubation, unbound Biotinylated anti-CRBN antibody and unbound Streptavidin-HRP are removed by washing, and two substrate solutions are added to the wells. colour develops in proportion to the amount of CRBN captured in each well. The colour development is stopped by addition of stop solution which changes the colour from blue to yellow and the intensity of the colour is then measured. The concentration of CRBN in the samples can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.

 : This is a ready to use ELISA kit that contains: Pre-coated 96 well microplate, standard solution, standard diluent, biotinylated detection antibody, steptavidin-HRP, wash buffer (25X), substrate solution A, substrate solution B, stop solution, plate sealers, foil pouch, and protocol manual.

Specifications

Specifications

  • Cdt:
    96 Tests
  • Désignation:
    Human CRBN ELISA kit
  • Type de test:
    Sandwich
  • Méthode de détection:
    Colorimetric
  • Primary antibody reactivity:
    Human
  • Portée de détection:
    1,5 - 24 ng/ml
  • Type déchantillon:
    Serum, plasma or other biological fluids
  • Time to Results:
    1 h 30 min
  • Température de stockage:
    4 °C
  • Cat. No.:
    ANTIA311905-96
  • Volume d'échantillon:
    40 μl
  • Format:
    Pre-coated
  • Cible:
    CRBN
  • Sensibilité:
    0,043 ng/ml
  • Durée du test:
    One step
  • Regulatory status:
    RUO
  • Durée de conservation:
    Store for 6 months at 4 °C

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.