Order Entry
Belgium
ContactUsLinkComponent
Human CCL25 ELISA Kit
Human CCL25 ELISA Kit
N° de catalogue ANTIA311091-96
Fournisseur:  ANTIBODIES.COM
New Product
Numéro CAS:  
undefined
Human CCL25 ELISA Kit
N° de catalogue ANTIA311091-96
Fournisseur:  ANTIBODIES.COM
New Product
Numéro CAS:  

Spécifications

  • Cdt:
    96 Tests
  • Durée du test:
    One step
  • Type de test:
    Sandwich
  • Format:
    Pre-coated
  • Primary antibody reactivity:
    Human
  • Cible:
    CCL25
  • Désignation:
    Human CCL25 ELISA kit
  • Type déchantillon:
    Serum, plasma or other biological fluids
  • Méthode de détection:
    Colorimetric
  • Time to Results:
    1 h 30 min
  • Durée de conservation:
    Store for 6 months at 4 °C
  • Portée de détection:
    50 - 800 ng/L
  • Température de stockage:
    4 °C
  • Volume d'échantillon:
    40 μl
  • Sensibilité:
    3,43 ng/L
  • Regulatory status:
    RUO

Spécifications

À propos de cet article

Human CCL25 ELISA kit is a 90 minute sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human CCL25 in serum, plasma, and other biological fluids.

  • Higher throughput: Get results in just 90 minutes, with a single wash step
  • Assay precision: Intra assay: CV <8%, Inter assay: CV <10%

Human CCL25 ELISA kit employs the sandwich enzyme immunoassay technique for the quantitative measurement of human CCL25 in serum, plasma or other biological fluids. An antibody specific for CCL25 has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the CCL25 present in each sample is bound to the wells by the immobilised antibody. Biotinylated anti-CCL25 antibody, which also binds the CCL25 present in each sample, and Streptavidin-HRP, which binds the Biotinylated anti-CCL25 antibody, are added and the microtiter plate is incubated. Following incubation, unbound Biotinylated anti-CCL25 antibody and unbound Streptavidin-HRP are removed by washing, and two substrate solutions are added to the wells. colour develops in proportion to the amount of CCL25 captured in each well. The colour development is stopped by addition of stop solution which changes the colour from blue to yellow and the intensity of the colour is then measured. The concentration of CCL25 in the samples can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.

Informations de commande: This is a ready to use ELISA kit that contains: Pre-coated 96 well microplate, positive control, negative control, HRP-conjugated detection antibody, sample diluent, substrate solution A, substrate solution B, stop solution, wash buffer (25X), plate sealers, foil pouch, and protocol manual.