Order Entry
Belgium
ContactUsLinkComponent
Human Tissue Factor ELISA Kit
Human Tissue Factor ELISA Kit
N° de catalogue ANTIA313339-96
Fournisseur:  ANTIBODIES.COM
New Product
Numéro CAS:  
undefined
Human Tissue Factor ELISA Kit
N° de catalogue ANTIA313339-96
Fournisseur:  ANTIBODIES.COM
New Product
Numéro CAS:  

Spécifications

  • Cdt:
    96 Tests
  • Durée du test:
    One step
  • Type de test:
    Sandwich
  • Format:
    Pre-coated
  • Hôte:
  • Primary antibody reactivity:
    Human
  • Cible:
    Tissue Factor
  • Désignation:
    Human Tissue factor ELISA kit
  • Environmentally Preferable:
  • Type déchantillon:
    Serum, plasma or other biological fluids
  • Méthode de détection:
    Colorimetric
  • Time to Results:
    1 h 30 min
  • Durée de conservation:
    Store for 6 months at 4 °C
  • Portée de détection:
    1,25 - 20 ng/ml
  • Température de stockage:
    4 °C
  • Volume d'échantillon:
    40 μl
  • Sensibilité:
    0,028 ng/ml
  • Regulatory status:
    RUO

Spécifications

À propos de cet article

Human Tissue Factor ELISA kit is a 90 minutes sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human Tissue Factor in serum, plasma, and other biological fluids.

  • Higher throughput: Get results in just 90 minutes, with a single wash step
  • Detection range: 1,25 to 20 ng/ml
  • Sensitivity: 0,028 ng/ml
  • Sample type: Serum, plasma or other biological fluids
  • Assay precision: Intra assay: CV <8%, Inter assay: CV <10%

Human Tissue Factor ELISA kit (A313339) employs the sandwich enzyme immunoassay technique for the quantitative measurement of human Tissue Factor in An antibody specific for Tissue Factor has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the Tissue Factor present in each sample is bound to the wells by the immobilised antibody. Biotinylated Anti-Tissue Factor antibody, which also binds the Tissue Factor present in each sample, and Streptavidin-HRP, which binds the Biotinylated Anti-Tissue Factor antibody, are added and the microtiter plate is incubated. Following incubation, unbound Biotinylated Anti-Tissue Factor antibody and unbound Streptavidin-HRP are removed by washing, and two substrate solutions are added to the wells. Colour develops in proportion to the amount of Tissue Factor captured in each well. The colour development is stopped by addition of stop solution which changes the colour from blue to yellow and the intensity of the colour is then measured. The concentration of Tissue Factor in the samples can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.

Informations de commande: This is a ready to use ELISA kit that contains: Pre-coated 96 well microplate, standard solution, standard diluent, biotinylated detection antibody, streptavidin-HRP, wash buffer (25X), substrate solution A, substrate solution B, stop solution, plate sealers, foil pouch, and protocol manual.