Order Entry
Belgium
ContactUsLinkComponent
Human UCN2/SRP ELISA Kit
Human UCN2/SRP ELISA Kit
Catalog # ANTIA78947-96
Supplier:  ANTIBODIES.COM
New Product
CAS Number:  
undefined
Human UCN2/SRP ELISA Kit
Catalog # ANTIA78947-96
Supplier:  ANTIBODIES.COM
New Product
CAS Number:  

Specifications

  • Cdt:
    96 Tests
  • Durée du test:
    Multiple steps
  • Type de test:
    Competitive
  • Format:
    Pre-coated
  • Primary antibody reactivity:
    Human
  • Cible:
    UCN2/SRP
  • Désignation:
    Human UCN2/SRP ELISA kit
  • Type déchantillon:
    Serum, plasma, tissue homogenates and other biological fluids
  • Réactivité croisée:
    Human UCN2/SRP ELISA Kit exhibits high specificity and excellent specificity for the detection of human UCN2/SRP. No significant cross-reactivity or interference between UCN2/SRP and analogues was observed.
  • Méthode de détection:
    Colorimetric
  • Time to Results:
    4 h 30 min
  • Principe du test:
    Quantitative
  • Durée de conservation:
    Store for 6 months at 4 °C
  • Portée de détection:
    31,25 - 2000 pg/ml
  • Température de stockage:
    4 °C
  • Volume d'échantillon:
    50 μl
  • Sensibilité:
    18,75 pg/ml
  • Regulatory status:
    RUO

Specifications

About this item

Human UCN2/SRP ELISA kit is a competitive Enzyme-Linked Immunosorbent Assay (cELISA) designed for the in vitro quantitative determination of human UCN2/SRP in serum, plasma, tissue homogenates, and other biological fluids.

  • Ready To Use ELISA Kit
  • Detection Range: 31,25 to 2000 pg/ml
  • Sensitivity: 18,75 pg/ml
  • Sample Type: Serum, plasma, tissue homogenates, and other biological fluids
  • Assay Precision: Intra - Assay: CV <8%, Inter - Assay: CV <10%

Human UCN2 / SRP ELISA Kit (A78947) employs the competitive enzyme immunoassay technique for the quantitative measurement of human UCN2 / SRP in serum, plasma, tissue homogenates, and other biological fluids. The 96-well microtiter plate has been pre-coated with UCN2 / SRP antigen. During the incubation, UCN2 / SRP present in the samples or standards competes with the fixed amount of immobilized UCN2 / SRP for binding sites on the Biotinylated Anti-UCN2 / SRP Antibody. The more UCN2 / SRP present in a sample or standard, the less Biotinylated Anti-UCN2 / SRP Antibody that binds to the plate. Following incubation, unbound Biotinylated Anti-UCN2 / SRP Antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualize the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is inversely proportional to the amount of UCN2 / SRP present in each sample or standard. The concentration of UCN2 / SRP can then be calculated by reading the O.D. absorbance at 450nm in a microplate reader and referring to the standard curve.

Ordering information: This is a ready to use ELISA kit that contains: Pre-coated 96 well microplate, lyophilised standard, sample dilution buffer, biotinylated detection antibody (100X), antibody dilution buffer, HRP-Streptavidin conjugate (SABC) (100X), SABC dilution buffer, TMB substrate, stop solution, wash buffer (25X), plate sealers, foil pouch, and protocol manual.