FAM Dye qPCR Calibration Plate *Optimised for ABI7500 Fast 96-Well*
AATB67006
New Product
- Couleur du socle:blanc
- Nb de puits:96
- Profil:Bas
- Jupe:Semi-skirted
- Couleur des puits:blanc
- Désignation:FAM Dye qPCR calibration plate
- Code-barres:Oui
- Cdt:1
The FAM dye qPCR calibration plate can be used to maintain the 7500 real-time PCR system with fast 96-well block.
- The qPCR calibration plate might significantly improve qPCR results with multiplexing by more accurately representing fluorescent spectra used in your real-time experiments
- This calibration plate is ready to use without any additional preparation steps required
For most qPCR instruments, the necessary calibrations should be run at least every six months.
: Avoid freeze-thawing plates. It is not recommended to use the plates more than 3 times. Plates can be stored for up to 12 months. Freeze (<−15 °C), and minimise light exposure.