Saisie de commande
Belgium
Lien pour commandesContactUsLinkComponent
Monkey IL-8 ELISA Kit

Monkey IL-8 ELISA Kit

Monkey IL-8 ELISA kit, 96 Tests
N° de catalogue ANTIA75544-96
Fournisseur:   Antibodies.com
Monkey IL-8 ELISA Kit
N° de catalogue ANTIA75544-96
Fournisseur:  Antibodies.com
Numéro CAS:  
Monkey IL-8 ELISA Kit
Monkey IL-8 ELISA Kit

Monkey IL-8 ELISA kit, 96 Tests

Numéro CAS:  
Prix
660,00 €
Connexion Pour la disponibilité
1 * 96 Tests
Product Details & Documents

Monkey IL-8 ELISA kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of monkey IL-8 in serum, plasma, tissue homogenates, and other biological fluids.

  • Ready to use ELISA kit
  • Detection range: 7,813 to 500 pg/ml
  • Sensitivity: 18,75 pg/ml
  • Sample type: Serum, plasma, tissue homogenates, and other biological fluids
  • Assay precision: Intra assay: CV <8%, Inter assay: CV <10%

Monkey IL-8 ELISA Kit (A75544) employs the sandwich enzyme immunoassay technique for the quantitative measurement of monkey IL-8 in serum, plasma, tissue homogenates, and other biological fluids. An antibody specific for IL-8 has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the IL-8 present in each sample is bound to the wells by the immobilised antibody. Following incubation, the wells are washed and then incubated with Biotinylated Anti-IL-8 Antibody, which binds the captured IL-8 present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualise the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of IL-8 captured in each well. The concentration of IL-8 can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.

Informations de commande: This is a ready to use ELISA kit that contains: Pre-coated 96 well microplate, lyophilised standard, sample dilution buffer, biotinylated detection antibody (100X), antibody dilution buffer, HRP-Streptavidin conjugate (SABC) (100X), SABC dilution buffer, TMB substrate, stop solution, wash buffer (25X), plate sealers, foil pouch, and protocol manual.

Spécifications

Spécifications

  • Cdt:
    96 Tests
  • Désignation:
    Monkey IL-8 ELISA kit
  • Type de test:
    Sandwich
  • Méthode de détection:
    Colorimetric
  • Primary antibody reactivity:
    Monkey
  • Principe du test:
    Quantitative
  • Portée de détection:
    7,813 - 500 pg/ml
  • Type déchantillon:
    Serum, plasma, tissue homogenates and other biological fluids
  • Réactivité croisée:
    Monkey IL-8 ELISA Kit exhibits high specificity and excellent specificity for the detection of monkey IL-8. No significant cross-reactivity or interference between IL-8 and analogues was observed.
  • Time to Results:
    4 h 30 min
  • Température de stockage:
    4 °C
  • Cat. No.:
    ANTIA75544-96
  • Volume d'échantillon:
    100 μl
  • Format:
    Pre-coated
  • Cible:
    IL-8
  • Sensibilité:
    18,75 pg/ml
  • Durée du test:
    Multiple steps
  • Regulatory status:
    RUO
  • Durée de conservation:
    Store for 6 months at 4 °C

Spécifications

Recommandations

Les recommandations seront personnalisées en fonction de vos préférences d’achat uniquement si vous avez donné votre consentement en activant l’option " Améliorer mon expérience d’achat " sur la page "Mon compte".

Dans le cas contraire, vous recevrez des recommandations génériques.