EPO Immunoaffinity Isolation Plate
STMC01640
:
- Cdt:1 KIT
- Hôte:
- Primary antibody reactivity:Human
- Cible:1640
- Environmentally Preferable:
Antibody-coated microtiter plate for immunoaffinity isolation of human EPO.
- Incorporates high-affinity monoclonal anti-EPO antibody (EPO-26 Antibody, Clone 26G9C10)
- Can be used to purify both endogenous and recombinant EPO from human serum, plasma, and urine samples
EPO Immunoaffinity Isolation Plate is a 96-well microtiter plate coated with a high-affinity monoclonal antibody (anti-EPO-26) against human erythropoietin (EPO). Each well can be used for immunoaffinity purification of natural or recombinant human EPO from serum, plasma, urine or other samples. EPO isolated using the EPO Immunaffinity Isolation Plate is appropriate for further analysis by methods such as SDS-PAGE, isoelectric focusing and mass spectrometry.
: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').