Saisie de commande
Belgium
Lien pour commandesContactUsLinkComponent
Mouse Cpeb2 ELISA Kit

Mouse Cpeb2 ELISA Kit

Mouse Cpeb2 ELISA kit, 96 Tests
N° de catalogue ANTIA312054-96
Fournisseur:   Antibodies.com
Mouse Cpeb2 ELISA Kit
N° de catalogue ANTIA312054-96
Fournisseur:  Antibodies.com
Numéro CAS:  
Mouse Cpeb2 ELISA Kit
Mouse Cpeb2 ELISA Kit

Mouse Cpeb2 ELISA kit, 96 Tests

Numéro CAS:  
Prix
565,00 €
Connexion Pour la disponibilité
1 * 96 Tests
Product Details & Documents

Mouse Cpeb2 ELISA kit is a 90 minutes sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of mouse Cpeb2 in serum, plasma, and other biological fluids.

  • Higher throughput: Get results in just 90 minutess, with a single wash step
  • Detection range: 125 to 2000 ng/L
  • Sensitivity: 9,02 ng/L
  • Sample Type: Serum, plasma or other biological fluids
  • Assay Precision: Intra-assay: CV <8%, Inter-assay: CV <10%

Mouse Cpeb2 ELISA kit employs the sandwich enzyme immunoassay technique for the quantitative measurement of mouse Cpeb2 in serum, plasma or other biological fluids. An antibody specific for Cpeb2 has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the Cpeb2 present in each sample is bound to the wells by the immobilised antibody. Biotinylated anti-Cpeb2 antibody, which also binds the Cpeb2 present in each sample, and Streptavidin-HRP, which binds the Biotinylated anti-Cpeb2 antibody, are added and the microtiter plate is incubated. Following incubation, unbound Biotinylated anti-Cpeb2 antibody and unbound Streptavidin-HRP are removed by washing, and two substrate solutions are added to the wells. Colour develops in proportion to the amount of Cpeb2 captured in each well. The colour development is stopped by addition of stop solution which changes the colour from blue to yellow and the intensity of the colour is then measured. The concentration of Cpeb2 in the samples can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.

Informations de commande: This is a ready-to-use ELISA kit that contains: Pre-coated 96 well microplate, standard solution, standard diluent, biotinylated detection antibody, steptavidin-HRP, wash buffer (25X), substrate solution A, substrate solution B, stop solution, plate sealers, foil pouch, and protocol manual.

Spécifications

Spécifications

  • Cdt:
    96 Tests
  • Désignation:
    Mouse Cpeb2 ELISA kit
  • Type de test:
    Sandwich
  • Méthode de détection:
    Colorimetric
  • Primary antibody reactivity:
    Mouse
  • Portée de détection:
    125 - 2000 ng/L
  • Type déchantillon:
    Serum, plasma or other biological fluids
  • Time to Results:
    1 h 30 min
  • Température de stockage:
    4 °C
  • Cat. No.:
    ANTIA312054-96
  • Volume d'échantillon:
    40 μl
  • Format:
    Pre-coated
  • Cible:
    Cpeb2
  • Sensibilité:
    9,02 ng/L
  • Durée du test:
    One step
  • Regulatory status:
    RUO
  • Durée de conservation:
    Store for 6 months at 4 °C

Spécifications