Multi-Use Cytotoxic Drug Spills Kit, Guest Medical
N° de catalogue GUESH9612
Fournisseur: GUEST
Spécifications
- Désignation:Multi-Use Cytotoxic Drug Spills Kit
- Cdt:1 KIT
Spécifications
À propos de cet article
The kit contains everything required for both liquid and powder spillages in one case that can be carried to the site where it is needed.
- 2 shakers of CT-Zorb Granules
- 3 Hazard Scoops and Scrapers
- 3 disposable tweezers
- 3 absorbent pads
- 3 sterile saline pods
- 6 detergent wipes
- 3 cytotoxic waste disposal bags
- 3 bag ties
- 3 pairs long cuff nitrile gloves
- 3 protective disposable gowns
- 3 pairs overshoes
- 3 FFP3 face masks
- Reusable goggles (BS EN 166 conformant)
- Warning sign
- Full instructions for use
The kit contains inert CT-Zorb Granules which will quickly and efficiently absorb all the moisture of a liquid spillage, thus containing the spill and preventing it spreading further. The granule-spill mixture can then be easily collected using the Guest Medical Scoops and Scrapers for disposal according to local procedure or regulations. Absorbent pads provide a simple method for collecting powder spillages when used with the sterile saline pods provided in the kit.