Specifications
- Nom de l'antigène:HLA-DR
- Clonalité:Monoclonal
- Clone:LN3
- Conjugaison:Unconjugated
- Hôte:Mouse
- Isotype:IgG2b kappa
- Réactivité:Rhesus monkey,Human
- ID de gène:3123
- Cdt:100 µG
Specifications
About this item
Mouse monoclonal IgG2b antibody against human, rhesus HLA-DR, unconjugated.
- B cells
- Dendritic cells
- Macrophages
- Monocytes
- Other
- T cells
The LN3 antibody reacts with the HLA-DR antigen, a major histocompatibility complex (MHC) class II receptor encoded within the human leukocyte antigen (HLA) complex on chromosome 6. HLA-DR is a heterodimeric transmembrane glycoprotein comprising non-covalently associated α (36 kDa) and β (27 kDa) subunits. MHC class II plays a central role in the presentation of antigen-derived peptides to CD4+ T cells, along with CD3/TCR and CD4. HLA-DR is primarily expressed on the surface of antigen-presenting cells, including B cells, dendritic cells, monocytes, macrophages, thymic epithelial cells, and activated T and natural (NK) cells. The LN3 antibody binds to an extracellular epitope that reportedly resides within the HLA-BR1 subunit of HLA-DR.
Type:
Antigen: HLA-DR
Clonality: Monoclonal
Clone: LN3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human;Rhesus monkey