Specifications
- Nom de l'antigène:Ym1
- Clonalité:Polyclonal
- Conjugaison:Unconjugated
- Hôte:Rabbit
- Isotype:IgG
- Réactivité:Mouse
- ID de gène:12655
- Taille:300 μl
- Cdt:0,3 mL
Specifications
About this item
Rabbit polyclonal IgG antibody against mouse Ym1, unconjugated.
Ym1 is an ~45 kDa secreted protein synthesized by activated mouse peritoneal macrophages during the inflammatory response elicited by parasitic infections. Ym1 is a granule protein in neutrophils and has been purified from eosinophilic crystals that form in the bronchoalveolar lavage fluid of mev/mev and SHIP (SH2-containing inositol 5’ phosphatase) knockout mice. The eosinophilic crystals formed in mice are considered to be histologically identical to the Charcot-Leyden crystals found in humans. Ym1 is homologous to T lymphocyte-derived eosinophil chemotactic factor (ECF-L) but elicits only a weak chemotactic response in eosinophils. Ym1 binds to heparin, GlcN oligomers and heparan sulfate. It belongs to a protein family related to chitinases but does not possess chitinase activity.
Type:
Antigen: Ym1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse