- Nom de l'antigène:SHIP2
- Clonalité:Polyclonal
- Conjugaison:Unconjugated
- Hôte:Rabbit
- Réactivité:Human,Mouse
- ID de gène:3636 (human), 16332 (mouse)
- Cdt:100 µl
Rabbit polyclonal antibody against human, mouse SHIP2, unconjugated.
- Granulocytes
- Subsets
- Lymphocytes
- Myeloid cells
SHIP2 is a 145 kDa SHIP-related protein expressed in both hematopoietic and nonhematopoietic cells. Like SH2-containing inositol phosphatase (SHIP), SHIP2 becomes tyrosine phosphorylated and associated with the adaptor protein, Shc, following cytokine, growth factor, chemokine, and immunoreceptor stimulation. SHIP2 also hydrolyzes the critical phosphatidylinositol (PI)-3-kinase (PI3K)-generated second messenger, PI-3,4,5-P3 (PIP3), to PI-3,4-P2 1,2 and therefore acts as an important negative regulator of the PI3K pathway. This antibody reacts with mouse and human full-length 145 kDa SHIP2 protein.
Type:
Antigen: SHIP2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse