Human Recombinant VEGF-121
Marketsource
Human Recombinant VEGF-121
Marketsource
Human Recombinant VEGF-121
Human Recombinant VEGF-121
Vascular endothelial growth factor (VEGF-121) is a naturally-occurring VEGF-A splice variant involved in embryonic vasculogenesis and angiogenesis. VEGF binds to VEGFR-1 and VEGFR-2, and activates Raf/MEK/ERK and PI3K/AKT pathways. VEGF-121 is released as a freely diffusible protein by a variety of normal and transformed cells. It plays an important role in neurogenesis both in vitro and in vivo (Storkebaum et al.). It has neurotrophic effects on neurons of the central nervous system and promotes growth and survival of dopaminergic neurons and astrocytes. VEGF also promotes growth and survival of vascular endothelial cells, monocyte chemotaxis, and colony formation by granulocyte-macrophage progenitor cells (Ferrara et al.).
 : Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
  • Catalog No:
  • STMC78127
  • STMC78127.1
  • STMC78127.2
  • Nom de la protéine/du peptide:
  • VEGF-121
  • VEGF-121
  • VEGF-121
  • Type de protéine/peptide:
  • Recombinant
  • Recombinant
  • Recombinant
  • Species:
  • Human
  • Human
  • Human
  • Purity:
  • ≥95%
  • ≥95%
  • ≥95%
Product Family Options
  • Cdt
  • 10 µG
    1 * 1 EA
    1 * 1 EA - 190,00 €
    190,00 €
    1 * 1 EA
       190,00 €
    Marketsource
    Specifications
    Cat. No.
    STMC78127
    Nom de la protéine/du peptide
    VEGF-121
    Type de protéine/peptide
    Recombinant
    Species
    Human
    Purity
    ≥95%
    Marketsource
  • 100 µG
    1 * 1 EA
    1 * 1 EA - 755,00 €
    755,00 €
    1 * 1 EA
       755,00 €
    Marketsource
    Specifications
    Cat. No.
    STMC78127.1
    Nom de la protéine/du peptide
    VEGF-121
    Type de protéine/peptide
    Recombinant
    Species
    Human
    Purity
    ≥95%
    Marketsource
  • 1.000 µG
    1 * 1 EA
    1 * 1 EA - 3 769,00 €
    3 769,00 €
    1 * 1 EA
       3 769,00 €
    Marketsource
    Specifications
    Cat. No.
    STMC78127.2
    Nom de la protéine/du peptide
    VEGF-121
    Type de protéine/peptide
    Recombinant
    Species
    Human
    Purity
    ≥95%
    Marketsource