Saisie de commande
Belgium
Lien pour commandesContactUsLinkComponent
CMV (UL44) Peptide Pool

CMV (UL44) Peptide Pool

1 AU
N° de catalogue STMC100-1405
Marketsource
Fournisseur:   Stemcell Technologies
CMV (UL44) Peptide Pool
N° de catalogue STMC100-1405
Fournisseur:  Stemcell Technologies
Marketsource
Numéro CAS:  
CMV (UL44) Peptide Pool
CMV (UL44) Peptide Pool

1 AU

Numéro CAS:  
Prix
Demander un prix
Connexion Pour la disponibilité
1 * 1 UO
Product Details & Documents

Stimulating T cells with CMV (UL44) peptide pool releases downstream cytokines and upregulates activation markers, enabling antigen-specific T cells to be detected or isolated for analysis. CMV (UL44) peptide pool is a lyophilised mixture of 106 peptides from DNA polymerase processivity factor UL44 of human cytomegalovirus (CMV; strain AD169), and consists of 15-mer peptides with 11-amino-acid overlaps that cover amino acids 1 to 433 on UL44. UL44 contains a nuclear localization signal (Alvisi et al.) and may bind DNA directly, or form a C clamp partially surrounding it, similar to proliferation cell nuclear antigen (PCNA) (Appleton et al.). UL44 has also been found to interact with viral (Kim and Ahn) and host (Strang et al.) proteins that contribute to viral DNA replication and efficient viral growth. Viral peptide pools are useful for a broad range of applications, including vaccine development, immunological research, and diagnostic assay development.

Attention: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').

Spécifications

Spécifications

  • Cdt:
    1 AU
  • Cat. No.:
    STMC100-1405
  • Purity:
    Average 70%
  • Nom de la protéine/du peptide:
    CMV (UL44) Peptide Pool

Spécifications