EasySep™ Buffer
Catalog # STMC100-0780
Supplier: STEMCELL Technologies
Specifications
- Désignation:Cell separation buffer for in vitro diagnostic (IVD) applications
- Cdt:1 L
Specifications
About this item
EasySep™ buffer is required for use with EasySep™ cell isolation kits that are in vitro diagnostic (IVD) medical devices for use with downstream diagnostic assays. EasySep™ buffer is an accessory to an in vitro diagnostic (IVD) medical device in Canada, Europe, the United Kingdom and the United States. EasySep™ buffer is indicated for in vitro diagnostic use by laboratory professionals.
Application: Cancer
Caution: The end user is responsible for determining whether the product is suitable for their specific application(s).
Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').