Gas Washing Bottle, Drechsel Pattern
Gas washing bottle, after Drechsel, without frit, 25 ml, 18,8/26
N° de catalogue NEUB05503000251906
Fournisseur: NEUBERT VOLUME GLASSWAERE
Gas Washing Bottle, Drechsel Pattern
N° de catalogue NEUB05503000251906
Fournisseur: NEUBERT VOLUME GLASSWAERE
Aller à :
Product Details & Documents
The bottle is made up of borosilicate 3.3 glass.
Complete with NS 29/32 and DIN 12596, including a safety attachment. For bottles up to 2 liters, the base height is 200 mm. Bottles exceeding 2 liters have a taller base height.
Spécifications
- RN:18,8/26
- Capacité:25 ml
- Désignation:Gas washing bottle, after Drechsel, without frit