Spécifications
- Désignation:Electrophoresis and visualisation package
- Cdt:1
Spécifications
À propos de cet article
Our best-selling M12 Complete™ just got even better. The M12 Ultra™ includes the M12 Complete™ electrophoresis package and the all-new TruBlu™ Jr. Blue light transilluminator. This dynamic duo enables real-time visualisation of SYBR® Safe or other blue light-reactive dye agarose gels.
- For blue light-reactive gels, overlay the orange filter on the M12 lid and slide the TruBlu™ Jr. underneath for instant visualisation
- The M12 Ultra™ offers the ultimate flexibility to either visualize gels in real-time
- The TruBlu™ Jr. can also be used to examine fluorescence from green fluorescent protein (GFP)
- Produces excellent results in 15 to 30 minutes
TruBlu™ Jr. Features:
Battery operated for convenient use in the classroom
Designed to work seamlessly with the EDVOTEK® M12 Electrophoresis System
Includes an orange contrast lid
Safe, non-UV wavelength
Blue Light emission spectrum centered around 470nm is compatible with SYBR® Safe, GelGreen® & other 10×6 cm viewing surface
M12 Complete™ Features:
Sleek new design speeds electrophoresis
Includes complete set of new and improved electrophoresis accessories
Now includes a DNA DuraGel™ (Cat. #S-43) for pipetting practice
Large color coded push tabs for easy lid insertion and removal
Pour spout for buffer disposal
Improved ventilation reduces lid condensation
User replaceable electrodes
Chamber reverse compatible with previous Edvotek® accessories